Description
- Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
- Corrosion resistant finish for wet or high humidity environments
- Antimicrobial is built into finish to keep product clean
Specifications
Specifications
| Product Type | Wire Shelf |
| Dimensions (L x W) | 24 x 24 in. |
| Finish | Metroseal 3 with Microban |