Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Metronidazole, Antiparasitic and Antibacterial Compound, Solid, 25 g

Catalog No. 502706790
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6790 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6790 Supplier Abcam Supplier No. AB14121825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nitroimidazole compound with antiparasitic and antibacterial activity and >99% purity.

  • Molecular weight: 171.15 Da
  • Purity: >99%
  • Description: Nitroimidazole compound with antiparasitic and antibacterial activity; active in vivo
  • Composition: 100% Metronidazole

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at 4°C.
Form Solid
Shipping Condition Ambient
Target Molecule Metronidazole
Quantity 25 g
For Use With (Application) Antiparasitic, Antibacterial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.