Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Metronidazole, Nitroimidazole Compound, Solid, 99% Purity, 5 g

Catalog No. 502706789
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6789 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6789 Supplier Abcam Supplier No. AB1412185G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nitroimidazole compound with antiparasitic and antibacterial activity and >99% purity.

  • Nitroimidazole compound with antiparasitic and antibacterial activity
  • Active in vivo
  • Purity: >99%
  • Molecular Weight: 171.15 Da
  • Composition: 100% Metronidazole
  • Form: Solid
  • CAS Number: 443-48-1

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Metronidazole
Shipping Condition Ambient
Alternate Part Number AB141218
Content And Storage Store at 4°C.
Form Solid
Quantity 5 g
For Use With (Application) Antiparasitic research, Antibacterial research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.