Learn More
Invitrogen™ METTL3 Monoclonal Antibody (OTI1B7)

Description
METTL3 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | METTL3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI1B7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | METTL3 |
| Gene Accession No. | Q86U44, Q8C3P7 |
| Gene Alias | 2310024F18Rik; adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase; fj82g07; hMETTL3; IME4; M6A; m6a methyltransferase; methyltransferase like 3; methyltransferase-like 3; methyltransferase-like protein 3; METTL3; mRNA (2'-O-methyladenosine-N(6)-)-methyltransferase; mRNA m(6)A methyltransferase; MTA70; MT-A70; N6-adenosine-methyltransferase 70 kDa subunit; N6-adenosine-methyltransferase catalytic subunit; N6-adenosine-methyltransferase subunit METTL3; Spo8; Unknown (protein for MGC:77093); wu:fj82g07; zgc:77093 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.