Learn More
Selleck Chemical LLC Metyrapone S5416-25mg
SDP
Supplier:  Selleck Chemical LLC S541625MG
Encompass_Preferred
Metyrapone (NSC-25265 SU-4885) is an inhibitor of the enzyme steroid 11-beta-monooxygenase (CYP11B1) that inhibits adrenal steroid synthesis
Catalog No. 50-311-0279
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.