Learn More
Lkt Laboratories Mevinolin, ≥97%, 50MG
SDP
Supplier:  Lkt Laboratories M168750MG
Encompass_Preferred
Statin, naturally produced by mushrooms; HMG-CoA reductase inhibitor. CAS: 75330-75-5. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. NC9125496
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.