Learn More
IBA LifeSciences MEXi-CM; 1000 ml
Supplier:  IBA LifeSciences 26010010
Encompass_Preferred
MEXi-CM culture medium is a serum-free, chemically defined medium especially developed for growth of high-density MEXi-293E cells. The low biotin content (120 µg/l) enables efficient and convenient purification with the aid of the Strep-tag technology.It is part of the MEXi mammalian expression system. The MEXi system was developed to offer an affordable mammalian protein expression system coming along with all essential components: • MEXi-293E cell line• A variety of pDSG expression vectors • MEXi-CM culture medium• MEXi-TM transfection mediumFurther advantages:• Transient expression for time- and cost-efficient protein production• Multiple expression vectors with different features• Optimized for Twin-Strep-tag:Strep-TactinXT system for highly pure proteins• IP-free system
Catalog No. 50-210-0163
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.