Learn More
Apexbio Technology LLC Lomerizine HCl 101477-54-7 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B17825.1
Encompass_Preferred
Lomerizine HCl (CAS 101477-54-7) is a small molecule antagonist of voltage-gated calcium channels exhibiting inhibitory activity on both L-type and T-type subtypes By modulating calcium influx across neuronal membranes lomerizine HCl is proposed to attenuate neuronal excitability associated with migraine pathophysiology This compound is utilized in biomedical research to investigate calcium channel function and the molecular mechanisms underlying migraine and related neurological disorders
Catalog No. 50-000-96495
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.