Learn More
Promega mFcgammaRIV ADCC Reporter Bioassay, Core Kit 5X, 1 each
Catalog No. PRM1215 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Each
Catalog No. Quantity
PRM1215 1 Each
1 options
Catalog No. PRM1215 Supplier Promega Supplier No. M1215
Add to Cart
Add to Cart
Request a Quote
The mFcgammaRIV ADCC Reporter Bioassay is a biologically relevant MOA-based assay that can be used to measure the activity of mouse antibodies that specifically bind and activate FcgammaRIV.
  • Measure Potency and Stability of Biologics that Bind and Activate mFcγRIV Receptor
  • Available in two kit formats:
  • Complete, everything you need to get started
  • Core, used with customer−defined Ab and target cells
Specifications
Content And Storage Components at different temps
Quantity 1 Each