Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MFN2 Monoclonal Antibody (N153/5), APC
GREENER_CHOICE

Catalog No. PIMA545669 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545669 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545669 Supplier Invitrogen™ Supplier No. MA545669
  • $508.00
    $466.65 / Each of 1

    Save $41.35

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Rat: 97% identity (226/231 amino acids identical). Human: 92% identity (214/231 amino acids identical) ~55% identity with Mitofusin-1. 1 μg/mL of MA5-45669 was sufficient for detection of Mitofusin-2 in 20 μg of mouse cardiac mitochondrial lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 90kDa. No cross-reactivity against Mitofusin-1. This antibody was formerly sold as clone S153-5.

This gene encodes a mitochondrial membrane protein that participates in mitochondrial fusion and contributes to the maintenance and operation of the mitochondrial network. This protein is involved in the regulation of vascular smooth muscle cell proliferation, and it may play a role in the pathophysiology of obesity. Mutations in this gene cause Charcot-Marie-Tooth disease type 2A2, and hereditary motor and sensory neuropathy VI, which are both disorders of the peripheral nervous system. Defects in this gene have also been associated with early-onset stroke. Two transcript variants encoding the same protein have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MFN2
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone N153/5
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene MFN2
Gene Accession No. O95140, Q80U63, Q8R500
Gene Alias CMT2A; CMT2A2; CPRP 1; CPRP1; D630023P19Rik; Fzo; HMSN6A; HSG; HSG protein; hyperplasia suppressor; hypertension related protein 1; hypertension-related protein; hypertension-related protein 1; Kiaa0214; LOW QUALITY PROTEIN: mitofusin-2; Marf; MFN 2; MFN2; mg:cb01g09; Mitochondrial assembly regulatory factor; mitochondrial transmembrane GTPase FZO1A; mitofusin 2; Mitofusin2; mitofusin-2; si:dkeyp-104h9.2; Transmembrane GTPase MFN2; wu:fb79a11
Gene Symbols MFN2
Host Species Mouse
Immunogen Fusion protein amino acids 370-600 (cytoplasmic N-terminus) of mouse Mitofusin-2.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170731, 64476, 9927
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.