Learn More
Cytiva MicroFunnel™ST Filter Funnel with GN-6 Membrane, 300 mL, 0.45 µm, White, Ungridded, Sterile, Bags of 5 Individual Bags (20/pkg)
Catalog No. 502063096 Shop All Cytiva Products
Encompass_Preferred
Convenient and Economical Choice for Sterility Testing Within Isolators-MicroFunnel ST filter funnels offer an alternative to costly closed-system sterility testing when using an isolator or containment suite-0.45 μm GN-6 Metricel membrane or low binding Supor membrane provides a choice for sterility testing applications in the pharmaceutical industry-Supor membrane is ideal for testing antibiotic solutions-Double bagged packaging ensures easy transfer through airlock-Simple squeeze separation design makes membrane retrieval easy-Meets requirements of U.S., Japan, and European Pharmacopeias for sterility testing This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3096 Supplier Cytiva Supplier No. 4812
Convenient and Economical Choice for Sterility Testing Within Isolators-MicroFunnel ST filter funnels offer an alternative to costly closed-system sterility testing when using an isolator or containment suite-0.45 μm GN-6 Metricel membrane or low binding Supor membrane provides a choice for sterility testing applications in the pharmaceutical industry-Supor membrane is ideal for testing antibiotic solutions-Double bagged packaging ensures easy transfer through airlock-Simple squeeze separation design makes membrane retrieval easy-Meets requirements of U.S., Japan, and European Pharmacopeias for sterility testing This product is not approved for use in medical, clinical, surgical or other patient applications.