Learn More
Ubpbio MG-132, 25MG
Supplier:  Ubpbio F1101
Encompass_Preferred
Inhibitors – MG-132 (UBPBio catalog# F1100, F1101, F1102) – MG-132 is a reversible proteasome inhibitor that belongs to the family known as peptide aldehydes. MG-132 is cell permeable and can be used to treat various mammalian cells to block proteasome activities. MG-132 can be dissolved in DMSO for stock solution up to 20 mM. The typical concentrations for cell culture use are 1-10 µM. For in vitro use, the typical concentrations are 50-100 µM. CAS#: 133407-82-6
Catalog No. 50-112-6890
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.