Learn More
MG 132, PROMO, Thermo Scientific Chemicals
Inhibitor of calpain proteases
Supplier: Thermo Scientific Chemicals J63250MAPR
Quantity | 10 mg |
---|
Description
Applications
MG 132 is used to inhibitor of calpain proteases, a proteasome and NF-kB inhibitor used as a tool for perturbing the proteasome-regulated degradation of intracellular proteins.
Solubility
Soluble in ethanol, chloroform, methanol, DMSO.
Notes
Protect from heat. Store away from oxidizing agents.

Specifications
133407-82-6 | |
Store at -20°C | |
475.63 g/mol | |
10 mg |
White | |
Crystalline Solid | |
MG 132 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.