Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam MG-132 Proteasome Inhibitor, Solid, >98% Purity, 25 mg

Catalog No. 502583379
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3379 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3379 Supplier Abcam Supplier No. AB14100325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly pure (greater than 98%) proteasome inhibitor for reliable research applications.

  • Molecular Weight: 475.6 Da
  • Purity: >98%
  • Form: Solid
  • Storage: Store at -20°C.
  • Shipping: Ambient - Can Ship with Ice
  • Composition: 100% MG 132

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Proteasome
Buffer 100% MG 132
Content And Storage Store at -20°C.
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Form Solid
Formulation 100% MG 132
Quantity 25 mg
For Use With (Application) Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular), Immunoprecipitation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.