Learn More
Medchemexpress LLC 2-(4-heptylphenethyl)-6-hydroxybenzoic acid | 1243583-85-8 | MFCD22123243 | 99.6% | 340.46 g/mol | C22H28O3 | 10 MG

Supplier: Medchemexpress LLC HY1588710MG
MG 149 is a small-molecule inhibitor of Tip60 (histone acetyltransferase) used in preclinical research to probe acetyltransferase activity and related mitochondrial pathways. It shows activity versus MOF/KAT8 and can influence PINK1-dependent mitochondrial clearance; supplied as a white to off-white solid with high analytical purity for laboratory use.
- Tip60 (HAT) inhibitor with reported IC50 ≈ 74 μM for Tip60 and ≈ 47 μM for MOF.
- High purity (HPLC 99.6%).
- Chemical formula C22H28O3; molecular weight 340.46 g/mol.
- White to off-white solid suitable for bench-scale research.
- Storage: powder at -20°C (stable 3 years) or 4°C (stable 2 years); in solvent at -80°C (2 years) or -20°C (1 year).
- Common applications: epigenetics assays, acetyltransferase inhibition studies, and mitochondrial/PINK1 pathway research.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.