Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius MGA Grade Glass Microfiber Filters w/o Binder

Catalog No. p-4340931 Shop All Sartorius Products
Change view
Click to view available options
Diameter (English):
4.92 in.
5.9 in.
7.2 in.
Diameter (Metric):
12.5 cm
15 cm
18.5 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Diameter (Metric)
14-555-182 4.92 in. 12.5 cm
14-555-180 7.2 in. 18.5 cm
14-555-181 5.9 in. 15 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-555-182 Supplier Sartorius Supplier No. FT31101125
Only null left
Add to Cart
Add to Cart

Binder free glass microfiber filters are suitable for analytical and gravimetric analyses. They are also used as prefilter.

Applications include: Clarification of buffer and reagent solutions, Filtration of algae cells.
TRUSTED_SUSTAINABILITY

Specifications

Air Resistance 36mbar
Color White
Filter Type Net Filter
Format Disc
For Use With (Equipment) Filtration Systems
Material Borosilicate Glass Microfiber
Particle Retention 1.6 μm
Sample Volume 435 mL
Sterility Non-sterile
Filtration Time 40 sec.
For Use With (Application) Clarification of buffer and reagent solutions
Grade MGA
Temperature (English) Max. 932°F
Temperature (Metric) Max 500°C
Thickness (English) 0.009 in.
Thickness (Metric) 0.25 mm
Type Glass Microfiber Filter
Diameter (English) 4.92 in.
Diameter (Metric) 12.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.