Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595338 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595338 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595338 Supplier Invitrogen™ Supplier No. PA595338
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: HELA whole cell, MCF-7 whole cell, SW620 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Functions as a dual-specificity transcription factor, regulating the expression of both MAX-network and T-box family target genes. Functions as a repressor or an activator. Binds to 5'-AATTTCACACCTAGGTGTGAAATT-3' core sequence and seems to regulate MYC-MAX target genes. Suppresses transcriptional activation by MYC and inhibits MYC-dependent cell transformation. Function activated by heterodimerization with MAX. This heterodimerization serves the dual function of both generating an E-box-binding heterodimer and simultaneously blocking interaction of a corepressor. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGA
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene MGA
Gene Accession No. Q8IWI9
Gene Alias AV312082; C130042M01Rik; C80739; D030062C11Rik; FLJ12634; KIAA0518; Kiaa4252; LOW QUALITY PROTEIN: MAX gene-associated protein; Mad5; maltase-glucoamylase (alpha-glucosidase); Max dimerization protein 5; MAX dimerization protein MGA; MAX gene associated; MAX gene-associated protein; MAX protein-associated protein-like protein; MAX-associated protein; MAX-interacting protein; MGA; MGA, MAX dimerization protein; MGA-like protein; MXD5; RGD1561597
Gene Symbols MGA
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human MGA (2376-2415aa QKEAEAFAYYRRTHTANERRRRGEMRDLFEKLKITLGLLH).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23269
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.