Learn More
MOLTOX Molecular Toxicology Inc Minimal Glucose Agar (MGA), 12 Well Plate, Bacto Agar, 3 mL/well, 2 plates/sleeve, Store at Room Temperature
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 2140S18.138
Encompass
Our Minimal Glucose Agars are specifically formulated for use in bacterial mutagenicity assays. Along with the standard formulation as described by Maron & Ames, Mutat. Res. 113:173, 1983, MOLTOX offers various other formulations such as histidine and biotin supplemented bottom agar. Our MGA""s are offered in case quantities or individual sleeves. MOLTOX provides all products necessary to conduct the Salmonella, E. coli, MOLTOX FT™, µAmes and UMU Mutagenicity Assays. All products are formulated and manufactured using the highest quality components and are consistent with the recommendations set forth by the historical literature corresponding to each test type. Certain Ames materials (e.g. STDiscs™ and Lyophilized S9) have been specifically developed in our laboratory for these assays. All products are thoroughly tested for performance in the assay and are accompanied by Quality Control and Formulation statements.
Catalog No. 50-948-653
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.