Learn More
Invitrogen™ MGARP Monoclonal Antibody (OTI5B6)

Description
MGARP Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | MGARP |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI5B6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MGARP |
| Gene Accession No. | Q8TDB4 |
| Gene Alias | C4orf49; CESP1; CESP-1; Corneal endothelium-specific protein 1; GS3582; HUMMR; hypoxia up-regulated mitochondrial movement regulator protein; MGARP; mitochondria localized glutamic acid rich protein; mitochondria-localized glutamic acid-rich protein; OSAP; ovary-specific acidic protein; Protein MGARP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.