Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGAT5B Rabbit anti-Human, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7207699 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1316 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1316 Supplier Proteintech Group Inc Supplier No. 169931AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Glycosyltransferase that acts on alpha-linked mannose of N-glycans and O-mannosyl glycans. Catalyzes the transfer of N-acetylglucosamine (GlcNAc) to the beta 1-6 linkage of the mannose residue of GlcNAcbeta1,2-Manalpha on both the alpha1,3- and alpha1,6-linked mannose arms in the core structure of N-glycan. Also acts on the GlcNAcbeta1,2-Manalpha1-Ser/Thr moiety, forming a 2,6-branched structure in brain O-mannosyl glycan. Plays an active role in modulating integrin and laminin-dependent adhesion and migration of neuronal cells via its activity in the O-mannosyl glycan pathway.

Specifications

Antigen MGAT5B
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MGAT5B
Gene Accession No. Q3V5L5
Gene Alias GlcNAc T Vb, GnT IX, GnT VB, hGnTVb, KIAA2008, MGAT5B
Gene Symbols MGAT5B
Host Species Rabbit
Immunogen MGAT5B Fusion Protein Ag10678
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146664, 303693
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.