Learn More
Thermo Scientific™ MGC DAU Control Sets
Description
Monitor Thermo Scientific™ CEDIA™ and Thermo Scientific™ DRI™ drugs of abuse testing assay performance with Thermo Scientific MGC DAU Control Sets. These controls contain high and low level controls at drug concentrations 25% below and above screening assay cutoffs to ensure quality laboratory results.
- Multi-constituent
- For use in the validation of drug of abuse assays in human urine
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | Amphetamine, Secobarbital, Nitrazepam, Benzoylecgonine, Benzoylecgonine, d-Methamphetamine, EDDP, Methadone Metabolite (EDDP), Morphine, Nitrazepam, Opiate 300 (Morphine), Secobarbital |
| Sample Type | Urine |
| Type | Optional DAU Control Set |
| Includes | Amphetamine: 225ng/mL and 375ng/mL; Barbiturate: 150ng/mL and 250ng/mL; Benzodiazepine: 150ng/mL and 250ng/mL; Cocaine: 225ng/mL and 375ng/mL; Methadone Metabolite: 75ng/mL and 125ng/mL; and Opiate 300: 225ng/mL and 375ng/mL |
| Quantity | 2 x 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.