Learn More
MGLL Mouse anti-Human, Clone: OTI4D1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503067

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Monoglyceride lipase functions together with hormone-sensitive lipase to hydrolyze intracellular triglyceride stores in adipocytes and other cells to fatty acids and glycerol. MGLL may also complement lipoprotein lipase in completing hydrolysis of monoglycerides resulting from degradation of lipoprotein triglycerides.Specifications
MGLL | |
Monoclonal | |
Unconjugated | |
MGLL | |
HU-K5, HUK5, MAGL, MGL | |
Mouse | |
Affinity Chromatography | |
RUO | |
11343 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI4D1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q99685 | |
MGLL | |
Full length human recombit protein of human MGLL produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.