Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mGluR7 Monoclonal Antibody (7G8D3)
GREENER_CHOICE

Catalog No. PIMA531754
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531754 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531754 Supplier Invitrogen™ Supplier No. MA531754
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31754 has been tested in indirect ELISA.

L-glutamate is the major excitatory neurotransmitter in the central nervous system, and it activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein-coupled receptors that have been divided into three groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties. Group I includes GRM1 and GRM5, and these receptors have been shown to activate phospholipase C. Group II includes GRM2 and GRM3, while Group III includes GRM4, GRM6, GRM7 and GRM8. Group II and III receptors are linked to the inhibition of the cyclic AMP cascade but differ in their agonist selectivities. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mGluR7
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7G8D3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene GRM7
Gene Accession No. Q14831
Gene Alias 6330570A01Rik; BB176677; C030018L03; E130018M02Rik; FLJ40498; G protein-coupled receptor GRM7; G protein-coupled receptor, family C, group 1, member G; GLUR7; glutamate metabotropic receptor 7; glutamate receptor; glutamate receptor, metabotropic 7; Gpr1g; Gprc1g; GRM7; Metabotropic glutamate receptor 7; metabotropic glutamate receptor mGluR7; metabotropic glutamate receptor subtype 7b; MGLU7; mGlu7a receptor; MGLUR7; PPP1R87; protein phosphatase 1, regulatory subunit 87
Gene Symbols GRM7
Host Species Mouse
Immunogen Purified recombinant fragment of human GRM7 (AA: 454-590) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2917
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.