Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGMT Recombinant Rabbit Monoclonal Antibody (JJ089-6)
GREENER_CHOICE

Catalog No. PIMA532461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532461 Supplier Invitrogen™ Supplier No. MA532461
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

O6-Methylguanine-DNA Methyltransferase (MGMT) is an important DNA repair protein involved in tumor cell resistance to the cytostatic activity of chemotherapeutic alkylating agents. This protein is also effective in protecting normal cells against the genotoxic and carcinogenic effects of DNA alkylation. The alkylating drug resistance is caused by MGMT's ability to remove DNA alkyl groups introduced in the O6 position of guanine. MGMT is expressed in highly variable amounts, depending upon the cell and tissue type, species, and cellular growth characteristics. In addition, MGMT activity varies among groups of tumors and within a particular type of tumor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGMT
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JJ089-6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene MGMT
Gene Accession No. P16455
Gene Alias 0-6-methylguanine-DNA methyltransferase; 6-O-methylguanine-DNA methyltransferase; Agat; AGT; AI267024; methylated DNA protein cysteine methyltransferase; methylated-DNA--protein-cysteine methyltransferase; methylguanine-DNA methyltransferase; MGMT; O(6)-alkylguanine-DNA alkyltransferase; O-6-alkylguanine-DNA alkyltransferase; O6-alkylguanine-DNA alkyltransferase; O-6-methylguan; O6-methylguanine-DNA methyltranferase; O-6-methylguanine-DNA methyltransferase; O6-methylguanine-DNA methyltransferase; O-6-methylguanine-DNA-alkyltransferase
Gene Symbols MGMT
Host Species Rabbit
Immunogen Recombinant protein within Human MGMT aa 1-140.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4255
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.