Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MGRN1 Monoclonal Antibody (OTI3C3)
GREENER_CHOICE

Catalog No. PIMA525499
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA525499 100 μL
1 options

Catalog No. PIMA525499

Supplier: Invitrogen™ MA525499

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MGRN1 Monoclonal Antibody for Western Blot, ICC/IF, Flow

Mahogunin (MGRN1) is a C3HC4 RING-containing protein with E3 ubiquitin ligase activity in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGRN1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3C3
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MGRN1
Gene Accession No. O60291
Gene Alias 2610042J20Rik; E3 ubiquitin-protein ligase MGRN1; KIAA0544; mahoganoid; mahogunin ring finger 1; mahogunin ring finger 1, E3 ubiquitin protein ligase; mahogunin RING finger protein 1; mahogunin, ring finger 1; mahogunin, ring finger 1a; md; mgrn1; mgrn1a; mKIAA0544; nc; probable E3 ubiquitin-protein ligase MGRN1; RGD1311862; RING finger protein 156; RING-type E3 ubiquitin transferase MGRN1; RNF156; wu:fi38c03; zgc:55978
Gene Symbols MGRN1
Host Species Mouse
Immunogen Full length human recombinant protein of MGRN1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23295
Target Species Human, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.