Learn More
Description
The MAPEG (Membrane Associated Proteins in Eicosanoid and Glutathione metabolism) family consists of six human proteins, two of which are involved in the production of leukotrienes and prostaglandin E, important mediators of inflammation. Other family members, demonstrating glutathione S-transferase and peroxidase activities, are involved in cellular defense against toxic, carcinogenic, and pharmacologically active electrophilic compounds. This gene encodes a protein that catalyzes the conjugation of glutathione to electrophiles and the reduction of lipid hydroperoxides. This protein is localized to the endoplasmic reticulum and outer mitochondrial membrane where it is thought to protect these membranes from oxidative stress. Four transcript variants of this gene encode one protein isoform.
- Theoretical MW: 42.79kDa
- Preparation Method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 Fast Flow
- Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
| For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
| Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
| Molecular Weight (g/mol) | 42.8 |
| Name | Human MGST1 Full-length ORF Recombinant Protein with GST-tag at N-terminal |
| pH Range | 8 |
| Preparation Method | In vitro wheat germ expression system |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE stained with Coomassie Blue |
| Quantity | 25 μg |
| Source | Wheat Germ (in vitro) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.