Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGST2 Antibody [Alexa Fluor™ Plus 488], Novus Biologicals™
SDP

Catalog No. RD10038817
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10038817 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10038817 Supplier R&D Systems Supplier No. NBP298515AFP488

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

MGST2 Polyclonal antibody specifically detects MGST2 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen MGST2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor Plus 488
Formulation 50mM Sodium Borate
Gene Alias EC 2.5.1.18, FLJ27438, GST2MGC14097, MGST-II, microsomal glutathione S-transferase 2, Microsomal GST-2, Microsomal GST-II, truncated microsomal glutathione S-transferase 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MGST2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4258
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.