Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MHC Class I (H-2Db) Monoclonal Antibody (28-14-8), Biotin, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. p-7069149
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-121-27 100 μg
50-121-28 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-121-27 Supplier Invitrogen™ Supplier No. 13599982
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: The 28-14-8 monoclonal antibody reacts with the mouse MHC class I, H-2D^b, and cross-reacts with H-2L^d, H-2D^q and/or H-2L^q. Binding of 28-14-8 to the alpha 3 domain of H-2L^d is not dependent on beta2-microglobulin. Applications Reported: 28-14-8 has been reported for use in flow cytometric analysis. Applications Tested: The 28-14-8 antibody has been tested by flow cytometric analysis of mouse splenocytes and can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.

This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC Class I (H-2Db)
Applications Flow Cytometry
Classification Monoclonal
Clone 28-14-8
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene H2-D1
Gene Accession No. P01895, P01896, P01897, P01899, P01900, P14426, P14427
Gene Alias H-2 class I histocompatibility antigen, D-B alpha chain; H-2D; H2-D; H-2D cell surface glycoprotein; H-2D(B); H2D1; H2-D1; H2DB; H2-DC1-beta transplantation antigen protein; H2L; H2LD; histocompatibility 2, D region locus 1; MHC class I antigen; MHC class I H2 antigen; MHC class I H-2Dp; MHC H2-D-q alpha-chain; Q5k protein
Gene Symbols H2-D1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14964
Target Species Mouse
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.