Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MHC Class I (H-2Kd/H-2Dd) Monoclonal Antibody (34-1-2S), eBioscience™, Invitrogen™

Catalog No. 5013443
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-134-43 500 μg
50-134-42 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-134-43 Supplier Invitrogen™ Supplier No. 14599885
  • $362.00
    $271.65 / Each of 1

    Save $90.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: The 34-1-2S monoclonal antibody reacts with the mouse MHC class I, H-2Kd and H-2Dd. This cytotoxic antibody also cross-reacts with Kb, s, r, q, p. Applications Reported: 34-1-2S has been reported for use in flow cytometric analysis, immunohistochemical staining of frozen mouse tissue sections, and complement-dependent cytotoxicity. Applications Tested: This 34-1-2S antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC Class I (H-2Kd/H-2Dd)
Applications Flow Cytometry
Classification Monoclonal
Clone 34-1-2S
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene H2-D1
Gene Accession No. P01895, P01896, P01897, P01899, P01900, P01901, P03991, P04223, P14426, P14427, P14428
Gene Alias class I major histocompatibility antigen H-2Kd; H-2 class I histocompatibility antigen, D-B alpha chain; H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H-2D; H2-D; H-2D cell surface glycoprotein; H-2D(B); H2-D1; H2-DC1-beta transplantation antigen protein; H2DD; H-2K; H2-K; H-2K(B); H-2K(d); H-2K(K); H-2K(Q); H2K1; H2-K1; H2KD; histocompatibility 2, D region locus 1; histocompatibility 2, K1, K region; K-f; MHC class I antigen; MHC class I H2 antigen; MHC class I H-2Dp; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I heavy chain H2-K; MHC H2-D-q alpha-chain; MHC H2-K transplantation antigen; Q5k protein
Gene Symbols H2-D1, H2-K1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14964, 14972
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.