Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MHC Class I Monoclonal Antibody (IVA26)
GREENER_CHOICE

Catalog No. PIMA544043 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544043 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544043 Supplier Invitrogen™ Supplier No. MA544043
  • $484.00
    $378.65 / Each of 1

    Save $105.35

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody recognizes an extracellular epitope on MHC class I (alpha + beta2m chains) expressed in various animals.

Like their mammalian counterparts, avian MHC Class I molecules, also known as B-F antigens, consist of a highly polymorphic alpha-chain noncovalently bound to the invariant beta2-microglobulin subunit. MHC Class I molecules are expressed on most nucleated cells where they present endogenously synthesized antigenic peptides to CD8+ T lymphocytes, which are usually cytotoxic T cells. The monoclonal antibody F21-2 also reacts with turkey MHC Class I. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC Class I
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone IVA26
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene HLA-A
Gene Accession No. P04439
Gene Alias A29 protein; alpha chain 2; antigen presentation; antigen presentation; BFIV major; BF12 major; antigen presenting molecule; Aw-33; Aw-74; B12; BF; B-F; B-F antigen; G-B1; B-F histocompatibility F10 antigen; B-F major; B-F minor; BF1; B-F1; BF12; BF2; BFa1; BFa2; B-F-beta-IV; B-FI; BFIV; B-FIV; BFIV21; B-F-S01; B-F-S02; B-F-S03; B-F-S04; B-F-S05; B-F-S06; B-F-S07; B-F-S09; B-F-S11; B-F-S12; B-F-S13; B-F-S14; BFw-03; BFw-05; BFz-01; B-LB2; BOLA class I histocompatibility antigen, alpha chain BL3-7-like; chicken MHC class I antigen; class I histocompatibility antigen, F10 alpha chain; EA1.2; EA2.1; F10 alpha chain; HFE; HLA A; HLA class I antigen; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA class I histocompatibility antigen, alpha chain E; HLA class I histocompatibility antigen, E alpha chain; HLA locus; HLA-6.2; HLAA; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-A11; HLA-A33; HLA-Aw33.1; HLA-DQB1; HLA-DRB1; HLAE; HLA-E; Human leukocyte antigen A; leukocyte antigen class I-A; lymphocyte antigen; major class I glycoprotein; Major histocompatibility complex class I antigen BF2; major histocompatibility complex class I glycoprotein; major histocompatibility complex class I glycoprotein haplotype B21; major histocompatibility complex, class I, A; major histocompatibility complex, class I, E; major histocompatibility complex, class I, G; MCH class I antigen; MHC; MHC BF1 class I; MHC BF2 class I; MHC class 1 antigen; MHC class I; MHC class I alpha chain 1; MHC class I alpha chain 2; MHC class I anti; MHC Class I antigen; MHC class I antigen A*30; MHC class I antigen A*33; MHC class I antigen A*74; MHC class I antigen alpha chain; MHC class I antigen B-F heavy chain; MHC class I antigen B-F major heavy chain; MHC class I antigen B-F minor heavy chain; MHC class I antigen E; MHC class I antigen heavy chain; MHC class I antigen HLA-A heavy chain; MHC class I antigen HLA-A33; MHC class I antigen null protein; MHC class I antigene; MHC class I B-F heavy chain; MHC class I glycoprotein; MHC class I heavy chain B-F; MHC class I molecule; MHC class I-alpha; MHC class II antigen; MHC class1 antigen; MHC classI antigen; MHC HLA-E alpha-1; MHC HLA-E alpha-2.1; minor antigen; minor class I glycoprotein; OLA-I; OVAR; Ovar-I; ovar-MHCI-B564; ovar-MHCI-D5; ovar-MHCI-F12; put. F10 preprotein (AA -22 to 323); QA1; sHLA-E; Soluble HLA class I histocompatibility antigen, alpha chain E; truncated MHC class I antigen
Gene Symbols HLA-A, OLA-I, OVAR
Host Species Mouse
Immunogen bovine lymphocytes.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100171394, 101108963, 3105
Target Species Human, Mouse, Rabbit, Bovine, Sheep, Pig, Avian
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.