Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MHC Class II (I-A/I-E) Monoclonal Antibody (M5/114.15.2), Brilliant Violet™ 421, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 404532182
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-453-2182 100 μg
40-453-2180 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-453-2182 Supplier Invitrogen™ Supplier No. 404532182
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

MHC (major histocompatibility complex) class II molecules are transmembrane glycoproteins expressed on the surface of professional antigen-presenting cells, such as macrophages, dendritic cells and B cells. Before their exposition on the cell surface, the MHC class II molecules react with endocytosed exogenous antigens, which are then presented to the T cells. The antigen-binding grove between MHC class II alpha and beta chain is open at both ends and is 15-24 amino acid residues long. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC Class II (I-A/I-E)
Applications Flow Cytometry
Classification Monoclonal
Clone M5/114.15.2
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 421
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene H2-Ab1
Gene Accession No. P01910, P01921, P04227, P04228, P04230, P06342, P06343, P06344, P06345, P06346, P14434, P14435, P14436, P14437, P14438, P14483, P23150
Gene Alias Abeta; AI323765; AI845868; cell surface glycoprotein; E-alpha-f; H-2 class II histocompatibility antigen, A beta chain; H-2 class II histocompatibility antigen, E-D alpha chain; H-2 class II histocompatibility antigen, E-K alpha chain; H-2 class II histocompatibility antigen, E-U alpha chain; H-2 class II histocompatibility antigen, I-E beta chain; H-2Ab; H2-Ab; H2-Ab1; H-2Ea; H2-Ea; H2-Ea-ps; H2Eb; H-2Eb; H2-Eb1; H2-iabeta; H2-IE-alpha; histocompatibility 2, class II antigen A, beta 1; histocompatibility 2, class II antigen E alpha; histocompatibility 2, class II antigen E alpha, pseudogene; histocompatibility 2, class II antigen E beta; Ia2; Ia-2; Ia3; Ia-3; Ia4; Ia-4; IAb; I-Ab; I-Abeta; I-Ad; I-Aq; I-E alpha MHC class II; I-E beta MHC class II; I-Ealpha; I-Ed; I-Ek; major histocompatibility complex class II beta chain; major histocompatibility complex H-2E beta chain; MHC class 2; MHC class II antigen A beta; MHC class II antigen E alpha; MHC class II H2-IA-beta-psi; MHC class II IE-b; MHC class II protein; MHC H2-IE-beta cell surface glycoprotein; response to metastatic cancers 1; Rmcs1
Gene Symbols H2-Aa, H2-Ab1, H2-Eb1
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14960, 14961, 14969
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.