Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MHC Class II (I-Ek) Monoclonal Antibody (14-4-4S), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. p-7069500
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-133-95 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-133-95 Supplier Invitrogen™ Supplier No. 14598082
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: The 14-4-4S monoclonal antibody reacts with the mouse and rat major histocompatibility complex class II, I-E subregion-encoded glycoprotein in mice strains of H-2^k, H-2^d, H-2^p and H-2^r. H-2^b, H-2^f and H-2^s haplotypes do not express the I-E antigen. MHC class II is present on B cells, monocytes, macrophages, and dendritic cells. The 14-4-4S mAb has been reported to block antigen presentation in in vitro assays. This antibody cross-reacts with rat. Applications Reported: 14-4-4S has been reported for use in flow cytometric analysis, immunoprecipitation, complement-dependent cytotoxity and immunohistochemical staining of frozen sections (Biotinylated 14-4-4S, cat. 13-5980, is recommended for IHC). Applications Tested: The 14-4-4S antibody has been tested by flow cytometric analysis of mouse splenocytes and can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

MHC (major histocompatibility complex) class II molecules are transmembrane glycoproteins expressed on the surface of professional antigen-presenting cells, such as macrophages, dendritic cells and B cells. Before their exposition on the cell surface, the MHC class II molecules react with endocytosed exogenous antigens, which are then presented to the T cells. The antigen-binding grove between MHC class II alpha and beta chain is open at both ends and is 15-24 amino acid residues long. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC Class II (I-Ek)
Applications Flow Cytometry
Classification Monoclonal
Clone 14-4-4S
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene H2-Eb1
Gene Accession No. P04230, P14439
Gene Alias AI323765; cell surface glycoprotein; E-alpha-f; H-2 class II histocompatibility antigen, E-D alpha chain; H-2 class II histocompatibility antigen, E-K alpha chain; H-2 class II histocompatibility antigen, E-U alpha chain; H-2 class II histocompatibility antigen, I-E beta chain; H-2Ea; H2-Ea; H2-Ea-ps; H2Eb; H-2Eb; H2-Eb1; H2-IE-alpha; histocompatibility 2, class II antigen E alpha; histocompatibility 2, class II antigen E alpha, pseudogene; histocompatibility 2, class II antigen E beta; Ia3; Ia-3; Ia4; Ia-4; I-E alpha MHC class II; I-E beta MHC class II; I-Ealpha; major histocompatibility complex H-2E beta chain; MGC117888; MHC class II antigen E alpha; MHC class II IE-b; MHC class II protein; MHC H2-IE-beta cell surface glycoprotein
Gene Symbols H2-Ea-ps, H2-Eb1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100504404, 14969
Target Species Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.