Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MHC Class II Monoclonal Antibody (YKIX334.2), FITC, eBioscience™
GREENER_CHOICE

Catalog No. p-7175680
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-2149 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-2149 Supplier Invitrogen™ Supplier No. 11590942
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: This YKIX334.2 monoclonal antibody reacts with canine major histocompatibility complex (MHC) Class II. The MHC plays a significant role in immunity by presenting self and non-self antigens to T cells. In dogs, MHC Class II is expressed on all T and B cells, macrophages, fibroblasts, and kidney interstitial cells. This monoclonal antibody has been reported to inhibit mixed lymphocyte reactions and T cell activation induced by MHC Class II antigen presentation in vitro. Also, YKIX334.2 recognition of MHC Class II is not allele-specific. Applications Reported: This YKIX334.2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This YKIX334.2 antibody has been pre-titrated and tested by flow cytometric analysis of canine peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

MHC (major histocompatibility complex) class II molecules are transmembrane glycoproteins expressed on the surface of professional antigen-presenting cells, such as macrophages, dendritic cells and B cells. Before their exposition on the cell surface, the MHC class II molecules react with endocytosed exogenous antigens, which are then presented to the T cells. The antigen-binding grove between MHC class II alpha and beta chain is open at both ends and is 15-24 amino acid residues long. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC Class II
Applications Flow Cytometry
Classification Monoclonal
Clone YKIX334.2
Concentration 5 μL/Test
Conjugate FITC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Alias beta 1 domain; B-L; B-L beta chain; B-L beta minor; B-LB; BLB major; BLB1; BLB2; B-LB2; B-LB21; B-LB21 major; BLB3; Bl-Beta II protein; B-LBII; class II histocompatibility antigen, B-L beta chain; class II histocompatibility antigen, M alpha chain; D6S222E; DMA; FECA-DRB; GSP-BLB1; GSP-BLB2; HLA class II histocompatibility antigen, DM alpha chain; HLADM; HLA-DMA; major histocompatibility class II antigen B-L beta; major histocompatibility complex class II B; Major histocompatibility complex class II beta chain BLB1; Major histocompatibility complex class II beta chain BLB1, (similar to HLA class II, D beta chain); major histocompatibility complex class II beta chain BLB1, (similar to HLA class II, D beta chain); Major histocompatibility complex class II beta chain BLB2, (similar to HLA class II, D beta chain); Major histocompatibility complex class II beta chain BLB3, (similar to HLA class II, D beta chain); major histocompatibility complex, class II, DM alpha; MHC 2; MHC Class II; MHC class II antigen; MHC class II antigen beta chain; MHC class II antigen B-F minor heavy chain; MHC class II antigen DMA; MHC class II antigen DRB; MHC Class II beta 1 and 2 domains; MHC class II beta 1 domain; MHC class II beta chain; MHC class II beta chain 1; MHC class II beta chain 2; MHC class II B-L beta chain; MHC II; MHCCII; MHCFECA-DRB; really interesting new gene 6 protein; RING6; S19; uncharacterized protein LOC101747454
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Canine
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.