Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MHC I Free Chain Monoclonal Antibody (A4), APC, eBioscience™
GREENER_CHOICE

Catalog No. 17995842
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-995-842 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-995-842 Supplier Invitrogen™ Supplier No. 17995842
  • $324.00
    $242.65 / Each of 1

    Save $81.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: The A4 monoclonal antibody reacts with the class I human leukocyte antigen (HLA) complex in the absence of beta2 microglobulin. There are three class I alpha-chain genes in humans, called HLA-A, -B and -C. Normally, HLA class I proteins are associated non-covalently with beta2 microglobulin (beta2M). The function of the HLA class I complex is to present peptides derived from intracellular pathogens, including viral pathogens, on the cell surface to antigen specific CD8+ cytotoxic T cells. Class I HLA is expressed by the majority of nucleated cells. Applications Reported: This A4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This A4 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

There are three class I alpha-chain genes in humans, called HLA-A, -B and -C. Normally, HLA class I proteins are associated non-covalently with B2 microglobulin (B2M). The function of the HLA class I complex is to present peptides derived from intracellular pathogens, including viral pathogens, on the cell surface to antigen specific CD8+ cytotoxic T cells. Class I HLA is expressed by the majority of nucleated cells. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC I Free Chain
Applications Flow Cytometry
Classification Monoclonal
Clone A4
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Alias MHC Free
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.