Learn More
Apexbio Technology LLC MI-2(Synonyms: Menin-MLL Inhibitor MI-2, MI2, MI 2, Menin inhibitor MI-2), 5mg, CAS: 1271738-62-5.
SDP
Supplier:  Apexbio Technology LLC A36025
Encompass_Preferred
MI-2 (CAS 1271738-62-5) is a small molecule inhibitor targeting the menin-MLL protein-protein interaction with an IC50 of 0 45 M The formation of menin-MLL fusion protein complexes resulting from chromosomal rearrangements plays an essential role in the pathogenesis of acute myeloid leukemia (AML) and acute lymphoblastic leukemia (ALL) MI-2 selectively binds wild-type menin disrupting its interaction with MLL-fusion proteins such as MLL-AF9 thereby inhibiting cellular proliferation and colony formation in MLL-AF9-transduced mouse bone marrow cells MI-2 serves as a research tool in studies of leukemia biology and menin-associated oncogenesis
Catalog No. 50-000-60822
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.