Learn More
Mi-2 Monoclonal Antibody (4D8), Active Motif™

Description
Specifications
Specifications
| Antigen | Mi-2 |
| Applications | Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 4D8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Gene Alias | 9530019N15Rik; AA617397; ATP-dependent helicase CHD4; BC005710; CHD4; CHD-4; chromodomain helicase DNA binding protein 4; Chromodomain-helicase-DNA-binding protein 4; D6Ertd380e; Mi-2; Mi-2 autoantigen; Mi-2 autoantigen 218 kDa protein; Mi-2 beta; MI2B; Mi-2b; Mi-2beta; Mi2-beta; mKIAA4075 |
| Host Species | Rat |
| Immunogen | This antibody was raised against a synthetic peptide corresponding to amino acid residues 1641-1662 of Drosophila Mi-2. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.