Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIB2/Skeletrophin Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501560328
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0328 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0328 Supplier Bethyl Laboratories, Inc Supplier No. A301414AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

E3 ubiquitin-protein ligase that mediates ubiquitination of Delta receptors, which act as ligands of Notch proteins. Positively regulates the Delta-mediated Notch signaling by ubiquitinating the intracellular domain of Delta, leading to endocytosis of Delta receptors.

Specifications

Antigen MIB2/Skeletrophin
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene MIB2
Gene Accession No. Q96AX9
Gene Alias MIB2, Mind Bomb-2, Novelzin, SKD, Skeletrophin, ZZANK1, ZZZ5, E3 ubiquitin-protein ligase MIB2, EC 6.3.2, EC 6.3.2.-, EC 6.3.2.13, FLJ20648, FLJ41406, Mind bomb homolog 2, mindbomb homolog 2 (Drosophila), Novel zinc finger protein, novelzin, Novelzin, Putative NF-kappa-B-activating protein 002N, SKD, Skeletrophin, Zinc finger ZZ type with ankyrin repeat domain protein 1, ZZ type with ankyrin repeat domain 1, ZZANK1
Gene Symbols MIB2
Host Species Rabbit
Immunogen Between 925 and 975
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 142678
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.