Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MICA/B Monoclonal Antibody (6D4), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5013154
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-131-54 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-131-54 Supplier Invitrogen™ Supplier No. 14578882
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: The 6D4 monoclonal antibody reacts with the human major histocompatibility complex (MHC) class I chain-related (MIC), MICA and MICB proteins. MICA and MICB are related proteins of 83% amino acid similarity, and show homology with classical human leukocyte antigen (HLA) molecules. The structure of MICA and MICB are similar to classical HLA class I chains, however they do not bind beta2 microglobulin or bind peptide typical of HLA class I. MICA and MICB are expressed on the cell surface of endothelial cells, fibroblasts, gastric epithelium and PHA-stimulated T cells, and act as a ligand for NKGD2 expressed on the surface of NK cells, gammadelta T cells and alpha beta CD8+ T cells. There is evidence to suggest that human cytomegalovirus (HCMV) subverts NK cell detection by inhibiting the function of MICB. Furthermore, MICA and MICB expression has been detected in several epithelial tumours isolated from breast, lung, ovary, prostate, colon and kidney. Applications Reported: This 6D4 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistochemical staining. (Please use Functional Grade purified 6D4, cat. 16-5788, in functional assays.). Applications Tested: This 6D4 antibody has been tested by flow cytometric analysis of HeLa cells. This can be used at less than or equal to 0.5 μg per test.

MICA encodes the higly polymorphic MHC (HLA) class I chain-related gene A. The protein product is expressed on the cell surface, although unlike canonical class I molecules does not seem to associate with beta-2-microglobulin. It is thought that MICA functions as a stress-induced antigen that is broadly recognized by intestinal epithelial gamma delta T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MICA/B
Applications Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 6D4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene MICA
Gene Accession No. Q29980, Q29983
Gene Alias HLA class I antigen; MHC class I chain-related protein A; MHC class I chain-related protein B; MHC class I mic-B antigen; MHC class I polypeptide-related sequence A; MHC class I polypeptide-related sequence B; MHC class I related sequence A; MHC class I-like molecule PERB11.2-IMX; MICA; MIC-A; MICB; MIC-B; PERB11.1; PERB11.2; sMICA; sMICB; soluble MICA; soluble MICB; stress inducible class I homolog; truncated MHC class I polypeptide-related sequence A
Gene Symbols MICA, MICB
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100507436, 4277
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.