Learn More
Leica Microsystems Mica WideFocal Live Cell Microhub Automated Microscope
Shop All Leica Microsystems ProductsDescription
Perform long-term live cell experiments up to 4x faster than traditional widefield systems, capturing events that may previously have been missed. Easily get quality images as Mica automatically corrects the focus during experiments. Minimise phototoxicity and maintain optimal cell health with full environmental control. Ideal for long-term and dynamic live cell studies of organoids, spheroids, and 3D cultures.
MICA BENEFITS:
- ACCESS FOR ALL: Mica requires minimal technical input and gives consistent results every time, so all lab members can leverage microscopy to make more discoveries.
- NO CONSTRAINTS: Visualize in widefield and switch effortlessly to confocal without ever moving your sample. Get true simultaneous capture of up to four labels without spatiotemporal mismatch at near-physiological conditions for live cells, or with fixed samples.
- RADICALLY SIMPLIFIED WORKFLOWS: Intelligent automation and AI-supported analysis of diverse samples, such as tissues on slides or well plates, enable greater efficiency and a faster track to publication.
Specifications
Specifications
| Type | Automated Imaging System |
| Camera | CMOS |