Learn More
Leica Microsystems Mica WideFocal Microhub Automated Microscope
Catalog No. 01930021 Shop All Leica Microsystems Products
Change view
Click to view available options
Camera:
CMOS
Catalog No. Camera
01-930-021 CMOS
1 options
Catalog No. 01-930-021 Supplier Leica Microsystems Supplier No. 111000033
Add to Cart
Add to Cart
Combine widefield and confocal imaging in an intuitive platform that simplifies advanced fluorescence imaging. Intelligent automation and AI-driven analysis streamline workflows, reduce complexity, and deliver high-resolution imaging for thick or complex samples.
MICA BENEFITS:
  • ACCESS FOR ALL: Mica requires minimal technical input and gives consistent results every time, so all lab members can leverage microscopy to make more discoveries.
  • NO CONSTRAINTS: Visualize in widefield and switch effortlessly to confocal without ever moving your sample. Get true simultaneous capture of up to four labels for fast acquisitions.
  • RADICALLY SIMPLIFIED WORKFLOWS: Intelligent automation and AI-supported analysis of diverse samples, such as tissues on slides or well plates, enable greater efficiency and a faster track to publication.
TRUSTED_SUSTAINABILITY
Specifications
Type Automated Imaging System
Camera CMOS