Learn More
MilliporeSigma™ Supelco™ Michel-Miller Column Plug Front Seal, PTFE
Catalog No. 111009352 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Thread Size:
80 mm
Catalog No. Thread Size
11-100-9352 80 mm
1 options
Catalog No. 11-100-9352 Supplier MilliporeSigma™ Supelco™ Supplier No. Z5071211EA
Add to Cart
Add to Cart
Solid plug for use as stopper with Ace-Threds. Seals M-M column ends, Plugs with No. 50 Ace-Thred fit Ace large columns.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
Thread Size 80 mm
Material PTFE