Learn More
MilliporeSigma™ Supelco™ Michel-Miller Swagelok™ adapter NPTF 1/16 in., PTFE
Catalog No. 111011024 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Thread Size:
Ace-Thred No. 11
Ace-Thred No. 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size
11-101-1024 Ace-Thred No. 11
11-101-0624 Ace-Thred No. 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-1024 Supplier MilliporeSigma™ Supelco™ Supplier No. Z5072371EA
Add to Cart
Add to Cart
Use to make Swagelok™ tubing connections with Ace threaded glassware. One end is Ace-Thred, other is NPTF. Connects tubing to M-M columns. Adapters with No. 50 Ace-Thred fit Ace large columns.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
Thread Size Ace-Thred No. 11
Material PTFE
Swagelok is a registered trademark of Swagelok Company