Learn More
MilliporeSigma™ Supelco™ Michel-Miller Swagelok™ adapter PTFE, NPTF 1/8 in.
Catalog No. 111010917 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Thread Size:
Ace-Thred No. 15
Catalog No. Thread Size
11-101-0917 Ace-Thred No. 15
1 options
Catalog No. 11-101-0917 Supplier MilliporeSigma™ Supelco™ Supplier No. Z5072881EA
Add to Cart
Add to Cart
Use to make Swagelok™ tubing connections with Ace threaded glassware. One end is Ace-Thred, other is NPTF. Connects tubing to M-M columns. Adapters with No. 50 Ace-Thred fit Ace large columns.
TRUSTED_SUSTAINABILITY
Specifications
Thread Size Ace-Thred No. 15
Material PTFE
Swagelok is a registered trademark of Swagelok Company