Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Miconazole Nitrate 98.0+%, TCI America™
SDP

Catalog No. M176925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M176925G 25 g
M17695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M176925G Supplier TCI America Supplier No. M176925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22832-87-7
Molecular Formula C18H15Cl4N3O4
Molecular Weight (g/mol) 479.14
MDL Number MFCD00058161
InChI Key MCCACAIVAXEFAL-UHFFFAOYNA-N
Synonym miconazole nitrate, albistat, andergin, aflorix, conofite, florid, micatin, gyno-monistat, epi-monistat, gyno-daktar
PubChem CID 68553
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole; nitric acid
SMILES O[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1

Specifications

Melting Point 184°C
Color White
Quantity 25 g
Formula Weight 479.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Miconazole Nitrate
RTECSNumber NI4771000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.