Learn More
Ohaus Micro Centrifuge PROMO
Catalog No. 01920170PM Shop All OHAUS Products
Change view
Click to view available options
Capacity:
18 x 1.5/2 mL
Catalog No. Capacity
01-920-170PM 18 x 1.5/2 mL
1 options
Catalog No. 01-920-170PM Supplier Ohaus Supplier No. 83041044PM
Get Up to 40% Off OHAUS Centrifuges
View all promotions
Add to Cart
Add to Cart
Frontier™ micro centrifuges deliver high-speed operation for a range of essential lab applications, such as DNA/RNA preparation, ultrafiltration, and more. Biocontainment rotors enable research involving hazardous samples.
Frontier™ micro centrifuge for a wide range of essential lab applications.
TRUSTED_SUSTAINABILITY
Specifications
Capacity 18 x 1.5/2 mL
Max. Speed 14,000 rpm
Max. RCF 16,000 x g
Refrigerated No
Product Type Microcentrifuge