Learn More
Azzota Corp Microporous Filter, Ptfe, Diameter: 25mm, Pore Size: 0.45um, 50/Pk
Supplier:  Azzota Corp BT250145
Encompass_Preferred
Microporous Filter, PTFE, Diameter: 25mm, Pore Size: 0.45um, 50/pk
Catalog No. 50-161-2788
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.