Learn More
Micro Particle Size Standard, Based on Polystyrene Monodisperse, Analytical Standard, Size: 4.0 m, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Physical Form | Particles |
| Concentration | 2% (solids) |
| MDL Number | MFCD00243243 |
| Synonym | Size standards |
| Quantity | 10 mL |
| Grade | Analytical Standard |
Coulter is a trademark of Beckman Coulter, Inc.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.