Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ 96-Well Reaction Tube/Tray/Retainer Set, 0.2 mL

Catalog No. 403086
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Assemblies
20 Assemblies
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-308-6 100 Assemblies
40-308-3 20 Assemblies
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-308-6 Supplier Applied Biosystems™ Supplier No. 403086
Add to Cart
Add to Cart

Includes

Contains 100 MicroAmp 96-Well Reaction Tube/Tray Retainer Sets. Store at room temperature.

Requires

Caps or full plate covers to seal tubes

Optimally designed for precise PCR on Applied Biosystems 0.2mL thermal cyclers.

  • Economical 100-unit bulk packs
  • Nonsterile and autoclavable
Greater Sample Uniformity
  • Use with all instruments that have heated sample covers to eliminate condensation and the need for a mineral oil overlay
  • Proprietary MicroAmp Reaction Tubes are essential for high-performance PCR and ensure the greatest temperature and amplification uniformity for samples

See user's manual or package insert for limited label license and trademark information.

PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 100 MicroAmp™ 96-Well Reaction Tube⁄Tray Retainer Sets. Store at room temperature.
Product Line MicroAmp
Color Natural
Product Type 96-Well Reaction Tube/Tray/Retainer Set
For Use With (Equipment) 2720 Thermal Cycler, 6100 Thermal Cycler, GeneAmp 9700, 2720 Thermal Cycler
Plate Compatibility 96-Well Plates
Quantity 100 Assemblies
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.