Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ 32-Well Clear Adhesive Film

Catalog No. A32812
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Films
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A32812 100 Films
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A32812 Supplier Applied Biosystems™ Supplier No. A32812
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with MicroAmp™ TriFlex 3 x 32-Well PCR Reaction Plate

  • Easy to apply and remove - simply peel off backing and apply firmly to 32-well plate
  • Provides an alternative to cap strips and seals 32-well samples securely to prevent sample evaporation and well-to-well contamination
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Polyethylene
No. of Wells 32
Quantity 100 Films
For Use With (Equipment) MicroAmp TriFlex 3 x 32-Well PCR Reaction Plate
Shipping Condition Room Temperature
Product Type Film, Seal, or Applicator
Product Line MicroAmp
Plate Compatibility 32-well Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.