Learn More
Cytiva Microarray Hybridization Solution v2.0
Catalog No. 45200580 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Piece
Catalog No. Quantity
45-200-580 1 Piece
1 options
Catalog No. 45-200-580 Supplier Cytiva Supplier No. RPK0325
Add to Cart
Add to Cart
Uses a blend of proprietary components to provide quality automated and manual slide hybridizations. Cytiva Microarray Hybridization Solution v2.0 features hybridization rate enhancers that improve the detection of rare targets, plus blocking agents that minimize nonspecific background.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Piece